missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, Reagent, ACS, Spectrum™ Chemical

BR132, 76-60-8, 62625-32-5, C21H14Br4O5S

$269.82 - $10202.57

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.01
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
IUPAC Name 3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
18601092
View Documents
Spectrum Chemical Mfg Cor
BR1325GM
5 g Amber Glass Bottle
Each for $269.82
Only null left
 
18601093
View Documents
Spectrum Chemical Mfg Cor
BR13225GM
25 g Amber Glass Bottle
Each for $1,070.64
Only null left
 
18601094
View Documents
Spectrum Chemical Mfg Cor
BR132100GM
100 g Amber Glass Bottle
Each for $3,601.32
Only null left
 
18601095
View Documents
Spectrum Chemical Mfg Cor
BR1321KGBL
1 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Bromocresol Green, Reagent, ACS , is a dye in the triphenylmethane family and is used as a tracking dye and pH indicator for DNA agarose gel electrophoresis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications

Chemical Identifiers

76-60-8
698.01
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
C21H14Br4O5S
FRPHFZCDPYBUAU-UHFFFAOYSA-N
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C

Specifications

76-60-8
C21H14Br4O5S
CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1C
698.01
Amber Glass Bottle
3.8 to 5.4
Bromocresol Green
100%
FRPHFZCDPYBUAU-UHFFFAOYSA-N
3,3-bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
ACS
Beige
5 g
Videos
SDS
Documents

Documents