missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol green indicator, Reag. Ph. Eur., Honeywell Fluka™
Reag. Ph. Eur.
$176.92 - $176.92
Chemical Identifiers
| CAS | 76-60-8 |
|---|---|
| Molecular Formula | C21H14Br4O5S |
| Molecular Weight (g/mol) | 698.014 |
| MDL Number | MFCD00005875 |
| InChI Key | FRPHFZCDPYBUAU-UHFFFAOYSA-N |
| Synonym | 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue |
| PubChem CID | 6451 |
| IUPAC Name | 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
| SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 76-60-8 | |
| 698.014 | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 6451 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br |
| C21H14Br4O5S | |
| MFCD00005875 | |
| 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
| 76-60-8 | |
| C21H14Br4O5S | |
| MFCD00005875 | |
| 3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br | |
| 698.014 | |
| 698.01 | |
| Glass Bottle | |
| Bromocresol green indicator, Reag. Ph. Eur. |
| 100.0000% | |
| C19H10Br4O5S | |
| 372527 | |
| FRPHFZCDPYBUAU-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
| 6451 | |
| Ph. Eur. | |
| 5 g |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
Recommended Storage : Room Temp