missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, ACS reagent

Catalog No. AC402930010
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402930010 1 g Glass bottle
AC402930050 5 g Glass bottle
AC402930100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC402930010 Supplier Thermo Scientific Chemicals Supplier No. 402930010

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
Synonym 3',3'',5',5''-Tetrabromo-m-cresolsulfonephthalein, BCG, 3', 3'', 5'
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

Melting Point 225.0°C
Clarity Passes Test
Merck Index 15, 1393
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethylacetate
Infrared Spectrum Authentic
Formula Weight 698.01
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.8 (yellow) to pH 5.4 (blue)
Color Beige to Brown
Quantity 1 g
Physical Form Powder
Chemical Name or Material Bromocresol Green
Show More Show Less
missing translation for 'healthHazard1' Warning
missing translation for 'healthHazard2' GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-972-8
RTECSNumber SJ7456000
TSCA TSCA

RUO – Research Use Only