missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green, ACS

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

$77.82 - $275.07

Chemical Identifiers

CAS 76-60-8
Molecular Formula C21H14Br4O5S
Molecular Weight (g/mol) 698.014
MDL Number MFCD00005874
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
PubChem CID 6451
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6522506
View Documents
Thermo Scientific Chemicals
J6522506
5 g
Each for $77.82
Only null left
 
AAJ6522514
View Documents
Thermo Scientific Chemicals
J6522514
25 g
Each for $275.07
Only null left
 
Description

Description

Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green is used in a variety of colorimetric detection technologies. It is used as a tracking dye for DNA agarose gel electrophoresis, in protein determinations and in charge-transfer complexation processes. In TLC, it is used for visualization of the compounds with functional groups whose pKa is below 5.0. And it is ready-to-use spray and dip reagent for chromatography.

Solubility
Soluble in water, ethanol, methyl cellosolve.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

76-60-8
698.014
FRPHFZCDPYBUAU-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
C21H14Br4O5S
MFCD00005874
6451
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br

Specifications

225°C (decomposition)
C21H14Br4O5S
372527
Soluble in water,ethanol,methyl cellosolve.
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
698.014
698.04
Brown to Yellow
Powder
76-60-8
MFCD00005874
14,1386
FRPHFZCDPYBUAU-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
6451
ACS
5 g
Bromocresol Green
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-972-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only