missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green

CAS: 76-60-8 | C21H14Br4O5S | 698.014 g/mol

Supplier:  Thermo Scientific Chemicals A1709006

 View more versions of this product

Catalog No. AAA1709006


Only null left

Description

Description

Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green is a pH sensitive indicator used in the growth medium for microorganisms and titrations. It is useful in various colorimetric detection studies and as an actively tracking dye for DNA agarose gel electrophoresis. It acts as an inhibitor of prostaglandin E2 transport protein. It is also used in TLC for visualizing compounds containing functional groups having pKa less than 5.

Solubility
Soluble in benzene, ethanol and diethyl ether. Sparingly soluble in water.

Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue).Incompatible with strong oxidizing agents.
Specifications

Specifications

Bromocresol Green
76-60-8
MFCD00005874
14,1386
Soluble in benzene,ethanol and diethyl ether. Sparingly soluble in water.
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
698.014
698.02
∼225°C (decomposition)
C21H14Br4O5S
372527
Bromcresol Green
FRPHFZCDPYBUAU-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
6451
5 g

Chemical Identifiers

76-60-8
698.014
FRPHFZCDPYBUAU-UHFFFAOYSA-N
6451
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
C21H14Br4O5S
MFCD00005874
Bromcresol Green
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-972-8

RTECSNumber : SJ7456000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only