missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, 0.1% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue, Certified, LabChem™

Supplier:  LabChem LC118501

 View more versions of this product

Catalog No. LC118501


Only null left

Description

Description

Specifications

Specifications

Bromocresol Green
62625-32-5 , 7732-18-5
C21H14Br4O5S·Na
Passes Test
Certified
1g/mL
1g/mL
500 mL
0.1% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue
99.9,0.1
Soluble in water
720.06
Poly Bottle
Carbon monoxide; Carbon dioxide; Nitrogen oxides; Hydrogen bromide
Green
Liquid

Chemical Identifiers

62625-32-5
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions