missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, 0.04% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue, Certified, LabChem™

$65.79 - $117.32

Chemical Identifiers

CAS 62625-32-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC118307
View Documents
LabChem
LC118307
125 mL Poly Bottle
Each for $65.79
Only null left
 
LC118301
View Documents
LabChem
LC118301
500 mL Poly Bottle
Each for $79.89
Only null left
 
LC118302
View Documents
LabChem
LC118302
1 L Poly Bottle
Each for $117.32
Only null left
 
Description

Description

Specifications

Chemical Identifiers

62625-32-5

Specifications

62625-32-5 , 7732-18-5
C21H14Br4O5S·Na
Passes Test
Certified
1g/mL
1g/mL
125 mL
Bromocresol Green, 0.04% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue
99.96,0.04
Soluble in water
720.06
Poly Bottle
Carbon monoxide; Carbon dioxide; Nitrogen oxides; Hydrogen bromide
Blue/Green
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents