missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 5443-49-2 |
|---|---|
| Molecular Formula | C9H7BrO |
| Molecular Weight (g/mol) | 211.06 |
| InChI Key | WQRWNOKNRHCLHV-TWGQIWQCSA-N |
| Synonym | alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo |
| PubChem CID | 5369403 |
| IUPAC Name | (Z)-2-bromo-3-phenylprop-2-enal |
| SMILES | C1=CC=C(C=C1)C=C(C=O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC171272500
|
N/A | 250g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 5443-49-2 | |
| 211.06 | |
| alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo | |
| (Z)-2-bromo-3-phenylprop-2-enal |
| C9H7BrO | |
| WQRWNOKNRHCLHV-TWGQIWQCSA-N | |
| 5369403 | |
| C1=CC=C(C=C1)C=C(C=O)Br |
Specifications
| 5443-49-2 | |
| 97.5% min. (GC) | |
| C6H5CH=C(Br)CHO | |
| alpha-bromocinnamaldehyde, 2-bromo-3-phenylacrylaldehyde, alphabrocine, 2-bromo-3-phenyl-2-propenal, bromocinnamal, alpha-bromocinnamic aldehyde, ccris 3924, b 37 van, 2-propenal, 2-bromo-3-phenyl, cinnamaldehyde, alpha-bromo | |
| C1=CC=C(C=C1)C=C(C=O)Br | |
| 211.06 | |
| 211.06 | |
| α-Bromocinnamaldehyde |
| Authentic | |
| C9H7BrO | |
| 07,358 | |
| WQRWNOKNRHCLHV-TWGQIWQCSA-N | |
| (Z)-2-bromo-3-phenylprop-2-enal | |
| 5369403 | |
| 98% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 226-637-6
RUO – Research Use Only