Learn More
R&D Systems™ BromoCatch™ Expression Plasmid (C-term, pCMV, HygB)
Catalog No. RDEHBC06050
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
RDEHBC06050 50 μg
1 options
Catalog No. RDEHBC06050 Supplier R&D Systems™ Supplier No. RDEHBC06050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This plasmid enables expression of proteins fused to the BromoCatchTM tag, a covalent self-labeling tag derived from a BET bromodomain.
Specifications
Quantity 50 μg