missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromobimane 98.0+%, TCI America™
$423.14 - $1271.29
Chemical Identifiers
| CAS | 71418-44-5 |
|---|---|
| Molecular Formula | C10H11BrN2O2 |
| Molecular Weight (g/mol) | 271.11 |
| MDL Number | MFCD00036951 |
| InChI Key | AHEWZZJEDQVLOP-UHFFFAOYSA-N |
| Synonym | Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione |
| PubChem CID | 114810 |
| ChEBI | CHEBI:62825 |
| IUPAC Name | 3-(bromomethyl)-2,5,6-trimethyl-1H,7H-[1,2]diazolo[1,2-a]pyrazole-1,7-dione |
| SMILES | CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B422020MG
|
TCI America
B422020MG |
20 mg |
Each for $423.14
|
|
|||||
|
B4220100MG
|
TCI America
B4220100MG |
100 mg |
Each for $1,271.29
|
|
|||||
Chemical Identifiers
| 71418-44-5 | |
| 271.11 | |
| AHEWZZJEDQVLOP-UHFFFAOYSA-N | |
| 114810 | |
| 3-(bromomethyl)-2,5,6-trimethyl-1H,7H-[1,2]diazolo[1,2-a]pyrazole-1,7-dione |
| C10H11BrN2O2 | |
| MFCD00036951 | |
| Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione | |
| CHEBI:62825 | |
| CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O |
Specifications
| 71418-44-5 | |
| Yellow | |
| MFCD00036951 | |
| Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione | |
| CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O | |
| 271.11 | |
| CHEBI:62825 | |
| ≥98.0% (HPLC) | |
| Bromobimane |
| 162°C | |
| C10H11BrN2O2 | |
| 20 mg | |
| AHEWZZJEDQVLOP-UHFFFAOYSA-N | |
| 3-(bromomethyl)-2,5,6-trimethyl-1H,7H-[1,2]diazolo[1,2-a]pyrazole-1,7-dione | |
| 114810 | |
| 271.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer