accessibility menu, dialog, popup

UserName

CAS RN 71418-44-5

H3CH3CONNCH3OBr

CAS RN 71418-44-5

IUPAC Name: 3-(bromomethyl)-2,5,6-trimethyl-1H,7H-[1,2]diazolo[1,2-a]pyrazole-1,7-dione
Synonyms: monobromobimane MBBR bromobimane bimane monobromide
Molecular Weight (g/mol): 271.11
Molecular Formula: C10H11BrN2O2
InChi Key: AHEWZZJEDQVLOP-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=O)N2N1C(CBr)=C(C)C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results