missing translation for 'onlineSavingsMsg'
Learn More

Bromobenzene 99.0+%, TCI America™

Catalog No. p-7118445
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B043925G 25 g
B0439500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B043925G Supplier TCI America Supplier No. B043925G
Only null left

Chemical Identifiers

CAS 108-86-1
Molecular Formula C6H5Br
Molecular Weight (g/mol) 157.01
MDL Number MFCD00000055
InChI Key QARVLSVVCXYDNA-UHFFFAOYSA-N
Synonym benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
PubChem CID 7961
ChEBI CHEBI:3179
IUPAC Name bromobenzene
SMILES BrC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 156°C
Quantity 25 g
UN Number 2514
Formula Weight 157.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromobenzene
EINECSNumber (3)-0032
RTECSNumber CY9000000
TSCA Yes