missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromoacetyl Bromide 98.0+%, TCI America™
Supplier: TCI America B0539500G
Specifications
| Bromoacetyl Bromide | |
| Yellow | |
| C2H2Br2O | |
| 500 g | |
| bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide | |
| C(C(=O)Br)Br | |
| 201.845 | |
| 201.85 | |
| Liquid |
| 598-21-0 | |
| 150°C | |
| MFCD00000115 | |
| 2513 | |
| LSTRKXWIZZZYAS-UHFFFAOYSA-N | |
| 2-bromoacetyl bromide | |
| 61147 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 598-21-0 | |
| 201.845 | |
| LSTRKXWIZZZYAS-UHFFFAOYSA-N | |
| 61147 | |
| C(C(=O)Br)Br |
| C2H2Br2O | |
| MFCD00000115 | |
| bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide | |
| 2-bromoacetyl bromide |
Safety and Handling
EINECSNumber : (9)-1233&(9)-1275
TSCA : Yes