missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl Bromide 98.0+%, TCI America™

Catalog No. p-7118508
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B053925G 25 g
B0539100G 100 g
B0539500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B053925G Supplier TCI America Supplier No. B053925G
Only null left

Chemical Identifiers

CAS 598-21-0
Molecular Formula C2H2Br2O
Molecular Weight (g/mol) 201.845
MDL Number MFCD00000115
InChI Key LSTRKXWIZZZYAS-UHFFFAOYSA-N
Synonym bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
PubChem CID 61147
IUPAC Name 2-bromoacetyl bromide
SMILES C(C(=O)Br)Br

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 g
UN Number 2513
Formula Weight 201.85
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bromoacetyl Bromide
EINECSNumber (9)-1233&(9)-1275
TSCA Yes