Learn More
Bromoacetonitrile, 97%
CAS: 590-17-0 | C2H2BrN | 119.95 g/mol
Supplier: Thermo Scientific Chemicals 221860250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bromoacetonitrile | |
| 590-17-0 | |
| 1.7200g/mL | |
| >110°C | |
| 96% min. (GC) | |
| C2H2BrN | |
| BrCH2CN | |
| 25 g | |
| 13,197 | |
| bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
| BrCC#N | |
| 119.95 | |
| 119.95 | |
| Liquid |
| 97% | |
| Colorless to Yellow | |
| 148°C to 150°C | |
| Authentic | |
| Glass bottle | |
| 1.4786 to 1.4806 | |
| MFCD00001884 | |
| 02, 216 | |
| 1.72 | |
| REXUYBKPWIPONM-UHFFFAOYSA-N | |
| 2-bromoacetonitrile | |
| 11534 | |
| 97% |
Chemical Identifiers
| 590-17-0 | |
| 119.95 | |
| REXUYBKPWIPONM-UHFFFAOYSA-N | |
| 11534 | |
| BrCC#N |
| C2H2BrN | |
| MFCD00001884 | |
| bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile | |
| 2-bromoacetonitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES:
GHS Signal Word: Danger
EINECSNumber : 209-672-1
RTECSNumber : AL7970000
RUO – Research Use Only