missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile, 97%

Catalog No. AC221861000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221861000 100 g Glass bottle
AC221860050 5 g Glass bottle
AC221860250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221861000 Supplier Thermo Scientific Chemicals Supplier No. 221861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 590-17-0 | C2H2BrN | 119.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N

Specifications

Color Colorless to Yellow
Density 1.7200g/mL
Boiling Point 148°C to 150°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4786 to 1.4806
Linear Formula BrCH2CN
Quantity 100 g
Beilstein 02, 216
Fieser 13,197
Specific Gravity 1.72
Formula Weight 119.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bromoacetonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES:
EINECSNumber 209-672-1
RTECSNumber AL7970000

RUO – Research Use Only