missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetonitrile, 96%

Catalog No. AAA1393309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1393309 10 g
AAA1393318 50 g
AAA1393330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1393309 Supplier Thermo Scientific Chemicals Supplier No. A1393309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 590-17-0 | C2H2BrN | 119.95 g/mol

Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrile
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N

Specifications

Density 1.722
Boiling Point 150°C
Flash Point 81°C (178°F)
Refractive Index 1.48
Quantity 10 g
UN Number UN3276
Beilstein 956569
Solubility Information Insoluble in water.
Formula Weight 119.95
Percent Purity 96%
Chemical Name or Material Bromoacetonitrile
Show More Show Less
Hazard Category H301+H311+H331-H314
Hazard Statement GHS H Statement
H301-H311-H331-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 209-672-1
RTECSNumber AL7970000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only