missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific™

Chemical Identifiers

CAS 42014-51-7
Molecular Formula C6H6BrNO4
Molecular Weight (g/mol) 236.02
MDL Number MFCD00058571
InChI Key NKUZQMZWTZAPSN-UHFFFAOYSA-N
Synonym n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
PubChem CID 3565210
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
SMILES C1CC(=O)N(C1=O)OC(=O)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427782500
SDS View Documents
Thermo Scientific Chemicals
427782500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427780010
SDS View Documents
Thermo Scientific Chemicals
427780010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427780050
SDS View Documents
Thermo Scientific Chemicals
427780050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

42014-51-7
236.02
NKUZQMZWTZAPSN-UHFFFAOYSA-N
3565210
C1CC(=O)N(C1=O)OC(=O)CBr
C6H6BrNO4
MFCD00058571
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate

Specifications

42014-51-7
Authentic
Glass bottle
MFCD00058571
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
C1CC(=O)N(C1=O)OC(=O)CBr
236.02
236.02
Bromoacetic acid N-hydroxysuccinimide ester
88% min.
90%
C6H6BrNO4
250mg
NKUZQMZWTZAPSN-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
3565210
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only