missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid N-hydroxysuccinimide ester, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 427780050

 View more versions of this product

Catalog No. AC427780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Bromoacetic acid N-hydroxysuccinimide ester
88% min.
90%
C6H6BrNO4
5g
NKUZQMZWTZAPSN-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
3565210
90%
42014-51-7
Authentic
Glass bottle
MFCD00058571
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
C1CC(=O)N(C1=O)OC(=O)CBr
236.02
236.02

Chemical Identifiers

42014-51-7
236.02
NKUZQMZWTZAPSN-UHFFFAOYSA-N
3565210
C1CC(=O)N(C1=O)OC(=O)CBr
C6H6BrNO4
MFCD00058571
n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
(2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only