Learn More
Bromoacetic acid, 98+%
CAS: 79-08-3 | C2H3BrO2 | 138.95 g/mol
$136.75 - $136.75
Chemical Identifiers
| CAS | 79-08-3 |
|---|---|
| Molecular Formula | C2H3BrO2 |
| Molecular Weight (g/mol) | 138.95 |
| MDL Number | MFCD00002678 |
| InChI Key | KDPAWGWELVVRCH-UHFFFAOYSA-N |
| Synonym | bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo |
| PubChem CID | 6227 |
| IUPAC Name | 2-bromoacetic acid |
| SMILES | C(C(=O)O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC106572500
|
Thermo Scientific Chemicals
106572500 |
250 g | Glass bottle |
Each for $136.75
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 79-08-3 | |
| 138.95 | |
| KDPAWGWELVVRCH-UHFFFAOYSA-N | |
| 6227 | |
| C(C(=O)O)Br |
| C2H3BrO2 | |
| MFCD00002678 | |
| bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
| 2-bromoacetic acid |
Specifications
| 79-08-3 | |
| White to Yellow | |
| 208.0°C | |
| Authentic | |
| Glass bottle | |
| BrCH2CO2H | |
| 250 g | |
| 1.93 | |
| bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
| KDPAWGWELVVRCH-UHFFFAOYSA-N | |
| 2-bromoacetic acid | |
| 6227 | |
| 99% | |
| Bromoacetic acid |
| 49.0°C to 51.0°C | |
| 1.9300g/mL | |
| 113°C | |
| 99% | |
| C2H3BrO2 | |
| MFCD00002678 | |
| 02,213 | |
| 15,1407 | |
| Solubility in water: 900g/100mL (25°C). Other solubilities:g/100g(25°C): methanol-900,ether-735,benzene-308,carbon tetrachloride-168,soluble in acetone and alcohol,practically insoluble in n-heptane | |
| C(C(=O)O)Br | |
| 138.95 | |
| 138.95 | |
| Crystalline Mass or Crystalline Powder, Flakes, Crystals and/or Chunks |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Danger
EINECSNumber : 201-175-8
RUO – Research Use Only