missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid, 98+%

CAS: 79-08-3 | C2H3BrO2 | 138.948 g/mol

Supplier:  Thermo Scientific Chemicals A144030I

 View more versions of this product

Catalog No. AAA144030I

Only null left
Explore available promotions

Description

Description

Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water, alcohol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to light and air. Handle and store under inert gas. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Bromoacetic acid
44°C to 49°C
207°C to 209°C
Pungent
BrCH2CO2H
5000 g
506167
14,1400
Soluble in water, alcohol
C(C(=O)O)Br
138.948
138.95
79-08-3
1.93
>110°C (230°F)
C2H3BrO2
MFCD00002678
UN3425
Hygroscopic; Light sensitive
bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
KDPAWGWELVVRCH-UHFFFAOYSA-N
2-bromoacetic acid
6227
≥98%

Chemical Identifiers

79-08-3
138.948
KDPAWGWELVVRCH-UHFFFAOYSA-N
6227
C(C(=O)O)Br
C2H3BrO2
MFCD00002678
bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo
2-bromoacetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301+H311+H331-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BROMOACETIC ACID, SOLID

EINECSNumber : 201-175-8

RTECSNumber : AF5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only