Learn More
Bromoacetic acid, 98+%
CAS: 79-08-3 | C2H3BrO2 | 138.948 g/mol
Supplier: Thermo Scientific Chemicals A144030B
Description
Bromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromoacetic acid is used in organic synthesis and as an alkylating agent. It is also used as a biochemical for proteomics research. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.
Solubility
Soluble in water, alcohol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to light and air. Handle and store under inert gas. Incompatible with strong oxidizing agents and strong bases.
Specifications
| Bromoacetic acid | |
| 44°C to 49°C | |
| 207°C to 209°C | |
| Pungent | |
| BrCH2CO2H | |
| 1000 g | |
| 506167 | |
| 14,1400 | |
| Soluble in water, alcohol | |
| C(C(=O)O)Br | |
| 138.948 | |
| 138.95 |
| 79-08-3 | |
| 1.93 | |
| >110°C (230°F) | |
| C2H3BrO2 | |
| MFCD00002678 | |
| UN3425 | |
| Hygroscopic; Light sensitive | |
| bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
| KDPAWGWELVVRCH-UHFFFAOYSA-N | |
| 2-bromoacetic acid | |
| 6227 | |
| ≥98% |
Chemical Identifiers
| 79-08-3 | |
| 138.948 | |
| KDPAWGWELVVRCH-UHFFFAOYSA-N | |
| 6227 | |
| C(C(=O)O)Br |
| C2H3BrO2 | |
| MFCD00002678 | |
| bromoacetic acid, monobromoacetic acid, bromoethanoic acid, acetic acid, bromo, to ntu, bromoacetate ion, acide bromacetique, 2-bromoacetyl group, 2-bromoethanoic acid, acetic acid, 2-bromo | |
| 2-bromoacetic acid |
Safety and Handling
GHS H Statement
H301-H311-H331-H314-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301+H311+H331-H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BROMOACETIC ACID, SOLID
EINECSNumber : 201-175-8
RTECSNumber : AF5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only