missing translation for 'onlineSavingsMsg'
Learn More

alpha'-Bromo-alpha,alpha,alpha-trifluoro-p-xylene, 98%

CAS: 402-49-3 | C8H6BrF3 | 239.04 g/mol

$153.98 - $153.98

Chemical Identifiers

CAS 402-49-3
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD00000403
InChI Key IKSNDOVDVVPSMA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
PubChem CID 123062
IUPAC Name 1-(bromomethyl)-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1CBr)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC312990050
SDS View Documents
Thermo Scientific Chemicals
312990050
5 g Glass bottle
Each for $153.98
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

402-49-3
239.04
IKSNDOVDVVPSMA-UHFFFAOYSA-N
123062
C1=CC(=CC=C1CBr)C(F)(F)F
C8H6BrF3
MFCD00000403
4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
1-(bromomethyl)-4-(trifluoromethyl)benzene

Specifications

402-49-3
100.0
White to Yellow
65°C to 69°C (5.0 mmHg)
Authentic
Glass bottle
CF3C6H4CH2Br
5 g
4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
C1=CC(=CC=C1CBr)C(F)(F)F
239.04
239.04
Crystalline Low Melting Solid
97.5
30°C to 34°C
1.5400g/mL
88°C
97.5% min. (GC)
C8H6BrF3
MFCD00000403
1.54
IKSNDOVDVVPSMA-UHFFFAOYSA-N
1-(bromomethyl)-4-(trifluoromethyl)benzene
123062
98%
α'-Bromo-α, α, α-trifluoro-p-xylene, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-947-8

SDS
Documents

Documents

RUO – Research Use Only