missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-p-tolunitrile, 98%

Catalog No. AC162760100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162760100 10 g Glass bottle
AC162760500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162760100 Supplier Thermo Scientific Chemicals Supplier No. 162760100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17201-43-3 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17201-43-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001829
InChI Key UMLFTCYAQPPZER-UHFFFAOYSA-N
Synonym 4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
PubChem CID 86996
IUPAC Name 4-(bromomethyl)benzonitrile
SMILES C1=CC(=CC=C1CBr)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 113°C to 117°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CN
Quantity 10 g
Beilstein 09, 499
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inert organic solvents
Formula Weight 196.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α-Bromo-p-tolunitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-246-0
TSCA TSCA

RUO – Research Use Only