missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-o-tolunitrile, 98%

Catalog No. AC163131000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163131000 100 g Glass bottle
AC163130050 5 g Glass bottle
AC163130250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC163131000 Supplier Thermo Scientific Chemicals Supplier No. 163131000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22115-41-9 | C8H6BrN | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22115-41-9
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00001794
InChI Key QGXNHCXKWFNKCG-UHFFFAOYSA-N
Synonym 2-bromomethyl benzonitrile, 2-cyanobenzyl bromide, alpha-bromo-o-tolunitrile, o-cyanobenzyl bromide, a-bromo-o-tolunitrile, alpha bromo-o-tolunitrile, 2 bromomethyl benzonitrile, 2-bromomethyl-benzonitrile, 2-cyanophenyl methyl bromide, benzonitrile, 2-bromomethyl
PubChem CID 89599
IUPAC Name 2-(bromomethyl)benzonitrile
SMILES BrCC1=CC=CC=C1C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Brown-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4CN
Quantity 100 g
Beilstein 09, 470
Formula Weight 196.05
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material α-Bromo-o-tolunitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only