missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%

Catalog No. AC146330050
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146330050 5 g Glass bottle
AC146330100 10 g Glass bottle
AC146330500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC146330050 Supplier Thermo Scientific Chemicals Supplier No. 146330050

CAS: 1765-40-8 | C7H2BrF5 | 260.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1765-40-8
MDL Number MFCD00000299
InChI Key XDEPVFFKOVDUNO-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
PubChem CID 74484
IUPAC Name 1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 19.0°C
Color Colorless to Yellow
Density 1.7200g/mL
Boiling Point 174.0°C to 175.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H2BrF5
Refractive Index 1.4700 to 1.4720
Linear Formula C6F5CH2Br
Quantity 5 g
Specific Gravity 1.72
Molecular Weight (g/mol) 260.989
Formula Weight 260.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material α-Bromo-2,3,4,5,6-pentafluorotoluene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only