missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 1M solution in acetic acid, AcroSeal™

Catalog No. p-3523380
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388401000 100 mL AcroSeal™ Glass bottle
AC388408000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC388401000 Supplier Thermo Scientific Chemicals Supplier No. 388401000
Only null left

CAS: 7726-95-6 | Br | 79.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Specifications

Chemical Name or Material Bromine
Color Red
Name Note 1M solution in acetic acid
CAS 7726-95-6, 64-19-7
Molecular Formula Br
Linear Formula Br2
MDL Number MFCD00010896
Concentration 0.90 to 1.10M
Merck Index 15, 1401
Density 3.111
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
Solubility Information Solubility in water: soluble.
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES [Br-]
IUPAC Name bromide
Molecular Weight (g/mol) 79.91
PubChem CID 24408
ChEBI CHEBI:29224
Formula Weight 159.82
Packaging AcroSeal™ Glass bottle
Quantity 100 mL
Flash Point 40°C
Physical Form Liquid
Show More Show Less

RUO – Research Use Only