Learn More
Brominated Vegetable Oil, FCC, Spectrum™ Chemical
V1031, 8016-94-2
$310.30 - $34920.18
Chemical Identifiers
CAS 8016-94-2
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601084
View Documents
Spectrum Chemical Mfg Cor
V1031500MLGL
500 mL Amber Glass Bottle
Each for $310.30
Only null left
18601085
View Documents
Spectrum Chemical Mfg Cor
V10314LTGL
4 L Amber Glass Bottle
Each for $1,056.06
Only null left
18601086
View Documents
Spectrum Chemical Mfg Cor
V103120LTBL
20 L Steel Can
Each for $4,763.36
Only null left
18601087
View Documents
Spectrum Chemical Mfg Cor
V1031200LTBL
200 L Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Spectrum™ Chemical Brominated Vegetable Oil, FCC , also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
8016-94-2
Specifications
8016-94-2
Amber Glass Bottle
1.325-1.335
100%
500 mL
FCC