missing translation for 'onlineSavingsMsg'
Learn More

Brominated Vegetable Oil, FCC, Spectrum™ Chemical

Catalog No. 18601084
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601084 500 mL Amber Glass Bottle
18601085 4 L Amber Glass Bottle
18601086 20 L Steel Can
18601087 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601084 Supplier Spectrum Chemical Mfg Cor Supplier No. V1031500MLGL
Only null left

V1031, 8016-94-2

Spectrum™ Chemical Brominated Vegetable Oil, FCC , also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8016-94-2

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1.325-1.335
Grade FCC