missing translation for 'onlineSavingsMsg'
Learn More

Bromide Standard for Ion Chromatography, 0.1M NaBr, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$327.38 - $327.38

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AS-BR9-5Y
View Documents
SPEX CertiPrep Group LLC
ASBR95Y
125 mL Triple-leached LDPE bottle
Each for $327.38
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Br-0.1M

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
sodium bromide
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004
[Na+].[Br-]

Specifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
Standard Solution
JHJLBTNAGRQEKS-UHFFFAOYSA-M
Water
102.89
253881
0.1M NaBr
7647-15-6,7732-18-5
BrNa
Ion Chromatography
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Certified Reference Material
[Na+].[Br-]
sodium bromide
125 mL
CHEBI:63004
Bromide Standard
Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only