missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine hydrochloride, 98+%

Catalog No. AAJ6345122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6345122 100 g
AAJ6345136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAJ6345122 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' J6345122

A mucolytic agent | CAS: 611-75-6 | C14H21Br2ClN2 | 412.59 g/mol

A mucolytic agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-75-6
Molecular Formula C14H21Br2ClN2
Molecular Weight (g/mol) 412.59
MDL Number MFCD00056626
InChI Key SXIVGYTWSVJOLM-UHFFFAOYNA-N
Synonym bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
PubChem CID 5702220
ChEBI CHEBI:31303
IUPAC Name hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
SMILES [H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N

Specifications

Melting Point 240°C to 244°C
Color White
Quantity 100 g
Beilstein 4848376
Merck Index 14,1391
Formula Weight 412.6
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Bromhexine hydrochloride
EINECSNumber 210-280-8
RTECSNumber XS9950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only