missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine hydrochloride, 98.0-102% (AT), MilliporeSigma™ Supelco™

Catalog No. 111014882
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014882 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014882 Supplier Merck Emd Millipore Supplier No. 51082100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 611-75-6
Physical Form Neat
Linear Formula C14H20 Br2N2 · HCl
MDL Number MFCD00056626
Synonym 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine hydrochloride; N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine hydrochloride; Bromohexine hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 412.59
Percent Purity 98.0 to 102% (AT)
Grade Analytical Standard