missing translation for 'onlineSavingsMsg'
Learn More

Bromfenac sodium, Thermo Scientific™

$265.48 - $891.40

Chemical Identifiers

CAS 91714-93-1
Molecular Formula C15H11BrNNaO3
Molecular Weight (g/mol) 356.15
MDL Number MFCD03701673
InChI Key HZFGMQJYAFHESD-UHFFFAOYSA-M
IUPAC Name sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate
SMILES [Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC468922500
View Documents
Thermo Scientific Chemicals
468922500
250 mg
Each for $265.48
Only null left
 
AC468920010
View Documents
Thermo Scientific Chemicals
468920010
1 g
Each for $891.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

91714-93-1
356.15
HZFGMQJYAFHESD-UHFFFAOYSA-M
[Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1
C15H11BrNNaO3
MFCD03701673
sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate

Specifications

Glass Bottle
91714-93-1
C15H11BrNNaO3
Yellow to Orange
[Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1
356.15
Bromfenac sodium
Available
Powder
MFCD03701673
HZFGMQJYAFHESD-UHFFFAOYSA-M
sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate
250 mg
Videos
Safety and Handling

Safety and Handling

Environment

Recommended Storage : Normal conditions

SDS
Documents

Documents