missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.500 N (N/2), 0.250 M as Br2, Ricca Chemical

$167.30 - $587.17

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
CAS Min % 92.0,4.7,1.28
CAS Max % 96.0,4.89,1.33
CAS 7758-01-2
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
117216
View Documents Promotion Details
Ricca Chemical Company
117216
500 mL Amber Glass Bottle
Each for $167.30
Only null left
 
1172-32
View Documents Promotion Details
Ricca Chemical Company
117232
1 L Amber Glass Bottle
Each for $277.67
Only null left
 
1172-1
View Documents Promotion Details
Ricca Chemical Company
11721
4 L Amber Glass Bottle
Each for $587.17
Only null left
 
R11720004F
View Documents
Ricca Chemical Company
R11720004F
4 L Cubitainer
Each for $318.53
Only null left
 
Description

Description

Specifications

Specifications

Bromate-Bromide Solution
92.0,4.7,1.28
7758-01-2
Liquid
Colorless
96.0,4.89,1.33
Laboratory
0.500Normal,0.250Molar as Br
Videos
SDS
Documents

Documents