missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.0500 N (N/20), Ricca Chemical

Catalog No. p-3251381
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
116832 1 L
11681 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 116832 Supplier Ricca Chemical Company Supplier No. 116832
Only null left

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
CAS 7732-18-5, 7758-01-2, 7758-01-2
CAS Min % 97.0, 0.97, 0.13
CAS Max % 100.0, 1.01, 0.14
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0500Normal
Show More Show Less