missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromate-Bromide Solution, 0.0200 N (N/50), Ricca Chemical
$133.93 - $256.47
Specifications
| Chemical Name or Material | Bromate-Bromide Solution |
|---|---|
| Color | Colorless |
| Name Note | 0.0200 N (N/50) |
| CAS | 1-2-7758 |
| Assay Percent Range | 0.02000 ± 0.00002 N at 20°C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R11640001C
|
Ricca Chemical Company
R11640001C |
1 L | Amber Glass Bottle |
Each for $133.93
|
|
||||
|
R11640004C
|
Ricca Chemical Company
R11640004C |
4 L | Amber Glass Bottle |
Each for $256.47
|
|
||||
Specifications
| Bromate-Bromide Solution | |
| 0.0200 N (N/50) | |
| 0.02000 ± 0.00002 N at 20°C | |
| Solubility in Water: Infinite | |
| Odorless | |
| Liquid |
| Colorless | |
| 1-2-7758 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Laboratory | |
| Aqueous Solution |