missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.0200 N (N/50), Ricca Chemical

Catalog No. R11640004C
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R11640004C 4 L Amber Glass Bottle
R11640001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R11640004C Supplier Ricca Chemical Company Supplier No. R11640004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
Name Note 0.0200 N (N/50)
CAS 1-2-7758
Assay Percent Range 0.02000 ± 0.00002 N at 20°C
Synonym dihydrogen oxide, dihydrogen monoxide
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Show More Show Less