missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromaminic acid, 90+%
CAS: 116-81-4 | C14H7BrNNaO5S | 404.17 g/mol
$70.08 - $197.71
Chemical Identifiers
| CAS | 116-81-4 |
|---|---|
| Molecular Formula | C14H7BrNNaO5S |
| Molecular Weight (g/mol) | 404.17 |
| MDL Number | MFCD00035694 |
| InChI Key | TXMRAEGWZZVGIH-UHFFFAOYSA-M |
| Synonym | bromaminic acid, bromamine acid, 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid, alizarine cyanol grey g, unii-sbz7fun4bk, 1-amino-4-bromoanthraquinone-2-sulphonic acid, 1-amino-4-bromoanthraquinone-2-sulfonic acid, sbz7fun4bk, 1-amino-4-bromo-9,10-dioxo-9,10-dihydro-anthracene-2-sulfonic acid, 2-anthracenesulfonic acid, 1-amino-4-bromo-9,10-dihydro-9,10-dioxo |
| PubChem CID | 22628 |
| IUPAC Name | 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonic acid |
| SMILES | [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3211014
|
Thermo Scientific Chemicals
H3211014 |
25 g |
Each for $70.08
|
|
|||||
|
AAH3211022
|
Thermo Scientific Chemicals
H3211022 |
100 g |
Each for $197.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 116-81-4 | |
| 404.17 | |
| TXMRAEGWZZVGIH-UHFFFAOYSA-M | |
| 22628 | |
| [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O |
| C14H7BrNNaO5S | |
| MFCD00035694 | |
| bromaminic acid, bromamine acid, 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid, alizarine cyanol grey g, unii-sbz7fun4bk, 1-amino-4-bromoanthraquinone-2-sulphonic acid, 1-amino-4-bromoanthraquinone-2-sulfonic acid, sbz7fun4bk, 1-amino-4-bromo-9,10-dioxo-9,10-dihydro-anthracene-2-sulfonic acid, 2-anthracenesulfonic acid, 1-amino-4-bromo-9,10-dihydro-9,10-dioxo | |
| 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonic acid |
Specifications
| 116-81-4 | |
| Odorless | |
| MFCD00035694 | |
| bromaminic acid, bromamine acid, 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid, alizarine cyanol grey g, unii-sbz7fun4bk, 1-amino-4-bromoanthraquinone-2-sulphonic acid, 1-amino-4-bromoanthraquinone-2-sulfonic acid, sbz7fun4bk, 1-amino-4-bromo-9,10-dioxo-9,10-dihydro-anthracene-2-sulfonic acid, 2-anthracenesulfonic acid, 1-amino-4-bromo-9,10-dihydro-9,10-dioxo | |
| [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O | |
| 404.17 | |
| 382.19 | |
| Bromaminic acid |
| ∽280°C (decomposition) | |
| C14H7BrNNaO5S | |
| 25 g | |
| TXMRAEGWZZVGIH-UHFFFAOYSA-M | |
| 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonic acid | |
| 22628 | |
| ≥90% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 204-159-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only