accessibility menu, dialog, popup

UserName

CAS RN 116-81-4

IUPAC Name: sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Synonyms: sodium 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonate
Molecular Weight (g/mol): 404.17
Molecular Formula: C14H7BrNNaO5S
InChi Key: TXMRAEGWZZVGIH-UHFFFAOYSA-M
SMILES: [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results