missing translation for 'onlineSavingsMsg'
Learn More

Bromaminic acid, 90+%

Catalog No. AAH3211022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3211022 100 g
AAH3211014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3211022 Supplier Thermo Scientific Chemicals Supplier No. H3211022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 116-81-4 | C14H7BrNNaO5S | 404.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 116-81-4
Molecular Formula C14H7BrNNaO5S
Molecular Weight (g/mol) 404.17
MDL Number MFCD00035694
InChI Key TXMRAEGWZZVGIH-UHFFFAOYSA-M
Synonym bromaminic acid, bromamine acid, 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid, alizarine cyanol grey g, unii-sbz7fun4bk, 1-amino-4-bromoanthraquinone-2-sulphonic acid, 1-amino-4-bromoanthraquinone-2-sulfonic acid, sbz7fun4bk, 1-amino-4-bromo-9,10-dioxo-9,10-dihydro-anthracene-2-sulfonic acid, 2-anthracenesulfonic acid, 1-amino-4-bromo-9,10-dihydro-9,10-dioxo
PubChem CID 22628
IUPAC Name 1-amino-4-bromo-9,10-dioxoanthracene-2-sulfonic acid
SMILES [Na+].NC1=C(C=C(Br)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O

Specifications

Melting Point ∽280°C (decomposition)
Odor Odorless
Quantity 100 g
Formula Weight 382.19
Percent Purity ≥90%
Chemical Name or Material Bromaminic acid
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 204-159-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only