missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRMS1 Recombinant Rabbit Monoclonal Antibody (23GB3015)
GREENER_CHOICE

Catalog No. MA553312
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553312 100 μL
MA553313 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553312 Supplier Invitrogen™ Supplier No. MA553312
Only null left

Rabbit Recombinant Monoclonal Antibody

BRMS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene reduces the metastatic potential, but not the tumorogenicity, of human breast cancer and melanoma cell lines. The protein encoded by this gene localizes primarily to the nucleus and is a component of the mSin3a family of histone deacetylase complexes (HDAC). The protein contains two coiled-coil motifs and several imperfect leucine zipper motifs. Alternative splicing results in two transcript variants encoding different isoforms.

Specifications

Antigen BRMS1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3015
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BRMS1
Gene Accession No. Q9HCU9
Gene Alias AV003220; AW554636; breast cancer metastasis suppressor 1; breast cancer metastasis-suppressor 1; Breast cancer metastasis-suppressor 1 homolog; BRMS 1; BRMS1
Gene Symbols BRMS1
Host Species Rabbit
Immunogen A synthesized peptide derived from BRMS1 (1-56AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25855
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less