missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Britton-Robinson Buffer, pH 7.00, adjusted to 0.1 M Ionic Strength, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Britton-Robinson Buffer |
| Color | Colorless Liquid |
| CAS | 7732-18-5, 1310-73-2, 7664-38-2, 7447-40-7, 10043-35-3, 64-19-7 |
| CAS Max % | 99, 0.26, 0.25, 0.18, 0.16, 0.15 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 7 |
| Quantity | 500 mL |
| Sterility | Non-sterile |