missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Britton-Robinson Buffer, pH 10.88, adjusted to 0.1 M Ionic Strength, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Britton-Robinson Buffer |
| Color | Colorless Liquid |
| CAS | 7732-18-5, 1310-73-2, 7664-38-2, 64-19-7, 10043-35-3 |
| CAS Max % | 99.18, 0.35, 0.21, 0.13, 0.13 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | 10.88 |
| Quantity | 500 mL |
| Sterility | Non-sterile |