missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brimonidine 98.0+%, TCI America™
Supplier: TCI America B4132200MG
Specifications
| Brimonidine | |
| 247°C | |
| MFCD00153878 | |
| 2811 | |
| XYLJNLCSTIOKRM-UHFFFAOYSA-N | |
| 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine | |
| 2435 | |
| 292.14 | |
| Crystalline Powder |
| 59803-98-4 | |
| C11H10BrN5 | |
| 200 mg | |
| brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline | |
| BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1 | |
| 292.14 | |
| CHEBI:3175 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 59803-98-4 | |
| 292.14 | |
| XYLJNLCSTIOKRM-UHFFFAOYSA-N | |
| 2435 | |
| 5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine |
| C11H10BrN5 | |
| MFCD00153878 | |
| brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline | |
| CHEBI:3175 | |
| BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1 |
Safety and Handling
RTECSNumber : VD1200000
TSCA : No