missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brimonidine, 98%

Supplier:  Thermo Scientific™ 461522500

 View more versions of this product

Catalog No. AC461522500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Brimonidine
0.3% max.
97.5% min. (HPLC)
MFCD00153878
brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
XYLJNLCSTIOKRM-UHFFFAOYSA-N
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
2435
292.13
59803-98-4
Authentic
C11H10BrN5
250mg
Solubility in water: insoluble.
BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1
292.14
CHEBI:3175
98%

Chemical Identifiers

59803-98-4
292.14
XYLJNLCSTIOKRM-UHFFFAOYSA-N
2435
5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
C11H10BrN5
MFCD00153878
brimonidine, bromoxidine, mirvaso, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl-6-quinoxalinamine, 5-bromo-n-4,5-dihydro-1h-imidazol-2-yl quinoxalin-6-amine, 5-bromo-6-2-imidazolin-2-ylamino quinoxaline, unii-e6gnx3hhte, brimonidine inn:ban, 3h brimonidine, 5-bromo-6-imidazolin-2-ylamino quinoxaline
CHEBI:3175
BrC1=C2N=CC=NC2=CC=C1NC1=NCCN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only