missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Yellow, Spectrum™ Chemical

$374.50 - $932.93

Chemical Identifiers

CAS 3051-11-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18609209
View Documents Promotion Details
N/A 25 g Amber Glass Bottle
Each for $374.50
Only null left
 
18609208
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $932.93
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3051-11-4

Specifications

3051-11-4
Reagent
Brilliant Yellow
1
Red
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.