missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Yellow, Spectrum™ Chemical

Catalog No. 18609209
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609209 25 g Amber Glass Bottle
18609208 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18609209 Supplier Spectrum Chemical Mfg Cor Supplier No. BR12525GM

Reagent grade Brilliant Yellow with an absorption maximum of 497 nm and a visual transition interval from pH 6.0 to pH 8.0.

  • Grade: Reagent
  • Molecular Formula: C26H18N4O8S2Na2
  • CAS Number: 3051-11-4
  • Absorption Max: 497 nm
  • Visual Transition Interval: pH 6.0 (yellow) to pH 8.0 (orange)

Chemical Identifiers

CAS 3051-11-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Color Red
pH 6 to 8
Quantity 25 g
Molecular Weight (g/mol) 624.56
Grade Reagent
Chemical Name or Material Brilliant Yellow

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.