missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Green, pure, high purity biological stain

CAS: 633-03-4 | C27H34N2O4S | 482.639 g/mol

$144.15 - $543.33

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
MDL Number MFCD00011880
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Basic Green 1, C.I. 42040, Ethyl Green
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC229601000
View Documents Promotion Details
Thermo Scientific Chemicals
229601000
100 g Glass bottle
Each for $144.15
Only null left
 
AC229605000
View Documents Promotion Details
Thermo Scientific Chemicals
229605000
500 g Glass bottle
Each for $543.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

633-03-4
482.639
NNBFNNNWANBMTI-UHFFFAOYSA-M
12449
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
C27H34N2O4S
MFCD00011880
Basic Green 1, C.I. 42040, Ethyl Green
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate

Specifications

1875 (at lambda min.)
633-03-4
C27H34N2O4S
13, 746
Basic Green 1, C.I. 42040, Ethyl Green
NNBFNNNWANBMTI-UHFFFAOYSA-M
Authentic
482.639
625 to 632nm (in 50:50 IMS:water)
10% max. (105°C)
Green
Brilliant Green, High purity biological stain
180.0°C
1 to 1.2
MFCD00011880
15, 1380
Solubility in water: 100g/L water (25°C). Other solubilities: soluble in alcohol
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
12449
482.64
Pure
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only