missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant Green, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 633-03-4 |
| Physical Form | Neat |
| Linear Formula | C27H34N2O4 S |
| MDL Number | MFCD00011880 |
| Melting Point | 210°C |
| Synonym | Astradiamant green GX; Basic Green 1; Diamond Green; Emerald Green; Ethyl Green; Malachite Green G; Solid Green JO |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 482.63 |
| Percent Purity | ≥98.0% (HPLC) |
| Show More |