missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Green

CAS: 633-03-4 | C27H34N2O4S | 482.639 g/mol

$73.99 - $140.49

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
MDL Number MFCD00011880
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Basic Green 1; C.I. 42040
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1280114
View Documents Promotion Details
Thermo Scientific Chemicals
A1280114
25 g
Each for $73.99
Only null left
 
AAA1280122
View Documents Promotion Details
Thermo Scientific Chemicals
A1280122
100 g
Each for $140.49
Only null left
 
Description

Description

Brilliant Green a stain used in bacteriological media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Green a stain used in bacteriological media.

Solubility
Soluble in water (100 mg/ml at 25°C), ethanol (50 mg/ml), methanol, and 2-methoxyethanol (60 mg/ml).

Notes
Store away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

633-03-4
482.639
NNBFNNNWANBMTI-UHFFFAOYSA-M
12449
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
C27H34N2O4S
MFCD00011880
Basic Green 1; C.I. 42040
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate

Specifications

210°C (decomposition)
C27H34N2O4S
3901207
Basic Green 1; C.I. 42040
NNBFNNNWANBMTI-UHFFFAOYSA-M
[4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
12449
Brilliant Green
633-03-4
MFCD00011880
14,1374
Soluble in water (100mg/ml at 25°C),ethanol (50mg/ml),methanol,and 2-methoxyethanol (60mg/ml).
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]
482.639
482.65
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 211-190-1

RTECSNumber : BP6825000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only